Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orbital Platform Shaker, PSU-10i, Grant Instruments

Supplier: Grant

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PSU-10I
89194-490EA 2550.31 USD InStock
89194-490
Orbital Platform Shaker, PSU-10i, Grant Instruments
Shakers and Mixers Orbital Shakers
Microprocessor controlled shaking platform providing smooth and quiet horizontal orbital motion for mixing in bottles, flasks and beakers

Variable speed control from 50 to 450rpm
Continuous or timer operation from 1 min to 96 h with automatic switch-off
Can be used at 5 to 45 °C, in an incubator or cold room
Can take loads up to 3kg and has an automatic load balancing system

Supplied without platform, required accessories must be ordered separately. Available with a choice of four interchangeable accessory platforms, to enable use with a wide range of vessels, including bottles, flasks and beakers, dishes, boxes and Petri dishes.

The shaker has an LCD display of both set and actual speed, plus set and elapsed time. An audible signal accompanies automatic switch-off to indicate the completion of a set time period. The soft start feature ensures a smooth ramp up of the shaking speed.

Certifications: CE marked.

Ordering information: Platform not included. Manufacturer's 2 year warranty.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty