Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Puritan PurFlock Ultra® Flocked Swabs, Puritan Medical Products
Supplier: Puritan Medical Products
PurFlock®
3706-U 25-3606-U 25-3406-U 3318-U 25-3506-U 3317-U 25-3806-U 25-3206-U 25-3316-U 25-3320-UEMB80 25-3306-U 3806-U 25-3318-U 3316-U 25-3317-U 3406-U
89497-308CS 561.74 USD InStock
89497-308 89194-880 89194-890 89194-884 89194-896 10124-680 76436-476 89194-893 89194-894 89194-888 89194-886 89194-898 75788-570 89428-648 10124-676 10124-674
Puritan PurFlock Ultra® Flocked Swabs, Puritan Medical Products
Swabs Swabs for Sampling
Engineered for purity and maximal cell yield with a small sample size
The uniquely designed fiber structure accounts for better cell yield in the specimen where sample size is limited.
Standard tip swabs are ideal for Buccall cell collection. Mini tip and Ultrafine tip swabs are best suited for Nasopharyngeal and male urethral cell collection. Micro Ultrafine tip swabs are ideal for pediatric applications. All swabs are made in the USA.
PurFlock Ultra® specimen collection devices are ideal for PCR, molecular assays, DFA testing, forensic applications, and direct antigen testing.
Certifications: CE marking for medical use. Patented: United States, Canada, Europe, Australia, Japan, and China. Patents Pending: Hong Kong.
Ordering information: Sterile swabs are individually wrapped. Nonsterile swabs are bulk packaged.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty