Would you like to log in again?
Multiple Polypropylene Bucket System, 5 Gallon (20 L), Contec®
Supplier: Contec
2757
89198-840CS 1439.11 USD InStock
89198-840
Multiple Polypropylene Bucket System, 5 Gallon (20 L), Contec®
Mops
The 5 Gallon Multiple Polypropylene Bucket System features a compact electropolished stainless steel rolling cart with two autoclavable polypropylene buckets. A third bucket is available for use when a three bucket system is required.
  • Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
  • Compatible with all Contec mops, depending upon wringer choice
  • System can be used with two or three buckets
  • Handle can be removed for storage
  • Compatible with most disinfectants
Compact design improves maneuverability and reduces storage space. The durable yet lightweight cart has four casters, two that lock easily with the touch of a foot. An extendable frame pulls out from the front of the cart to accommodate the third bucket. Includes a stainless steel connecting kit to prevent drips between the buckets.
The cart features a unique removable handle for easier autoclaving and storage. For maximum flexibility, we offer two types of wringers that are ordered separately. Choose from a stainless steel downpress wringer, or a stainless steel wringer that fits directly on the waste bucket.
Ordering information: Wringer options must be ordered separately. See the Wringers, Contec® product page for compatible options.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty