ES653-95
89198-918CS
204.2
USD
InStock
89198-918
EXCEL Alcohol 95% for histology
EXCEL Alcohol
Azer EXCEL alcohol is designed to prevent over-dehydration in tissue samples
- Formulation does not use ethanol
- Ideal for small specimens and biopsies
- No Biopsy Sample-Drying Ethanol with Excel
EXCEL alcohol is a blend of isopropyl and methyl alcohols. Because it does not use ethanol, it will prevent overdehyrdation of small specimens and biopsy samples.