Would you like to log in again?
GL Thread Caps/Pouring Rings, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7622-37 7622-155 7622-32 7622-58 7622-56 7622-114 7622-24 7622-02 7622-06 7622-35 7622-52 7622-21 7622-60 7622-103 7622-121 7622-14 7622-38 7622-65 7622-107 7622-54 7622-64 7622-04 7622-62 7622-124
89199-092CS 135.8 USD InStock
89199-092 89199-094 89059-808 89059-806 89059-804 89059-802 89059-800 89083-182 89059-788 89059-798 89083-184 89059-786 89059-796 89083-186 89083-178 89059-784 89059-794 89083-188 89059-792 89199-096 89059-790 89199-098 89199-100 89083-180
GL Thread Caps/Pouring Rings, Ace Glass Incorporated
Closures Bottle Closures
These screw caps and pouring rings are for use with GL threaded caps
Red caps (except GL120 cap) have a PTFE liner, all others are with lip seal. For all GL threaded bottles.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty