Would you like to log in again?
Cap, Premium, Ace Glass Incorporated
Supplier: Ace Glass
7627-03 7627-02
89199-106CS 289.84 USD InStock
89199-106 89199-108
Cap, Premium, Ace Glass Incorporated
Closures Bottle Closures
This premium cap and pour ring are designed for GL45 thread Duran bottles
Made of colorless, TPCh260 resin so that there are no leachables. Cap has high chemical resistance, and is leak-tight due to a PTFE coated silicone seal. Meets USP standards. Autoclavable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty