Would you like to log in again?
Cap/Pour Ring, GL Threaded Bottles, Ace Glass Incorporated
Supplier: Ace Glass
7628-03 7628-04
89199-110CS 697.43 USD InStock
89199-110 89199-112
Cap/Pour Ring, GL Threaded Bottles, Ace Glass Incorporated
Closures Bottle Closures
These GL threaded caps have quick-release closure for the new GL80 thread, wide-mouth bottles
Made of white polyarylsulphone 1 (PSU) resin, which provides high thermal, mechanical, and chemical compatibility. Inner seal is coated both sides with PTFE. PSU pour ring also available. The Maximum Temperature for 89199-110 is +180°C and N/A for the 89199-112.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty