Would you like to log in again?
Tissue-Tek® Accu-Edge® Grossing Stations, Sakura Finetek
Supplier: Sakura
Tissue-Tek®
108 106 105 107
89199-654EA 43565.94 USD InStock
89199-654 89199-656 89199-650 89199-652
Tissue-Tek® Accu-Edge® Grossing Stations, Sakura Finetek
Furniture Casework
Accu-Edge® Grossing Stations offer the opportunity to customize a grossing station to fit any laboratory's specific requirements
  • Standard grossing station features include:
  • 1/2 HP disposal
  • Dissecting area rinse
  • Polyethylene dissecting board
  • Formalin container with spigot
  • GFCI waterproof electrical receptacles
  • Halogen light on flex arm
  • Magnetic instrument holder
  • Large sink with mixing faucet
  • Dual recessed halogen lights
  • Small specimen sink with cold water valve and basket
  • Spray hose assembly
  • C-fold stainless paper towel holder
  • Vacuum breaker protection
  • Exhaust duct to building
Signed blueprints provided with signature required by facilities.
Ordering Information: For a detailed listed of optional accessory items and workstation components, see Tissue-Tek® Accu-Edge® Pathology Station Accessories, Sakura Finetek.
Constructed from high-quality stainless steel, the Tissue-Tek® Accu-Edge® Grossing Station is a floor model that stands at counter height. These pathology grossing stations are available in elevating or non-elevating formats. Elevating stations offer the ability to elevate from 32" to 44" to accommodate all lab personnel. Stations are availabe with or without hands-free infrared controls for hot, cold, rinse, and disposal functions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty