Would you like to log in again?
ScrubKITS™, Corrosion Cleaning Kits, Foamtec International
Supplier: Foamtec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ScrubKITS™
HT4500-SSC2 HT4500-DM2 HT4500-DM1
89176-720CS 3184.77 USD InStock
89176-720 89200-068 89176-718
ScrubKITS™, Corrosion Cleaning Kits, Foamtec International
Wipes Cleaning Kits and Pads
High precision remediation kits ease cleaning of corroded, rusted, or rouged 316 or 304 Stainless Steel.
Kits contain aggressive, medium, fine and super fine ScrubPADS, UltraSOLV® Sponges and MiraWIPE® wipers which enable operators to return rusted 304 and 316 stainless steel to a like-new condition using a fast process.
Starter kits include cleaning and polishing step-by-step instructions and training video for the removal of corrosion and preventing re-rusting. Refill kits provide larger quantities of ScrubPADS, UltraSOLV® Sponges and MiraWIPES® only.
Kits are compatible for use in ISO Class 5 Cleanrooms, and allow for remediation using DI or WIFI water in place of harsh chemicals, which enables the process to be done outside of shutdowns.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty