Would you like to log in again?
Disposable Automation Reservoirs, Thermo Scientific
Supplier: Thermo Scientific
1064-15-8 1064-15-6 1064-05-8 1064-05-6 1064-15-7 1064-05-7
89200-078CS 237.09 USD InStock
89200-078 89200-088 89200-084 89200-086 89200-080 89200-082
Disposable Automation Reservoirs, Thermo Scientific
Reagent Reservoirs
Disposable Automation Reservoirs are of polypropylene construction and are designed for simple, economical handling of reagents on automated systems
The standard microplate dimensions enables reservoirs to be used in plate handlers or stackers.
With the same dimensions as standard microplates, these disposable reservoirs can be used in plate handlers or stackers. The removable splash baffles prevent reagents from splashing out when being moved on robotic decks. They are compatible with virtually any automated liquid handler as well as all of our automated pipette tips.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty