Would you like to log in again?
Sample Tubes for Roche Cobas Analysers, Simport Scientific
Supplier: Simport
B721-1
89200-108CS 451.91 USD InStock
89200-108
Sample Tubes for Roche Cobas Analysers, Simport Scientific
Tubes Test Tubes
Graduated sample cup is for use on the Cobas® Analyzer 6000
This graduated false bottom tube has a capacity of 2.5mL and will accept bar code labels.
Made of clear polystyrene to allow visualization of contents
Ordering information: Caps can be ordered separately, see seris T404-3.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty