Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Corning® Gosselin™ Autoclave Bags, Corning

Supplier: Corning
CoolCell® Corning®
SA33-10 SA14-10 SA09-01 SA03-01 SA09-10 SA14-01 SA03-10 SA21-01 SA33-01 SA21-10
75799-270CS 267.84 USD InStock
75799-270 75799-258 89200-848 89200-856 75799-264 89200-844 89200-852 75799-260 89200-840 75799-262
Corning® Gosselin™ Autoclave Bags, Corning
Bags Sterilization Pouches
These autoclave bags are made of PP.

  • Every bag is 40 µm thick
  • Thermal resistance of 134 °C
  • Wide range of volumes available (3000 to 33000 ml)
  • Nonsterile

Bags must not be tightly sealed to withstand the temperature recommended for 20 minutes in autoclave.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty