Would you like to log in again?
Corning® Gosselin™ Straight Containers, Corning
Supplier: Corning
Corning®
FL100-05 FL60-04 FL300-04 FL52-02
89200-908CS 287.91 USD InStock
89200-908 89200-918 89200-910 89200-912
Corning® Gosselin™ Straight Containers, Corning
Sample Containers General Purpose Containers
Made of PP, natural or colored.
  • Available in sterile or under aseptic conditions packed versions
  • Very good chemical and temperature resistance
  • Graduated
  • Without cap
Ideal for sampling, storage and transport of liquid and solid samples.
Sized to provide adequate volume for milk sampling. It is shock resistant and is assured sterile 10-3.
Corning® Gosselin™ is a Corning brand of laboratory plasticware designed for use in industrial microbiology labs such as Food and Beverage, Environmental/Water Testing, etc.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty