Would you like to log in again?
Steril-Peel® Self-Seal Sterilization Pouches, GS Medical Packaging
Supplier: GS Medical Packaging
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
222565 222100 222104 222555 222816 222101
89201-574CS 532.81 USD InStock
89201-574 89201-572 89201-571 89201-578 89201-576 89201-568
Steril-Peel® Self-Seal Sterilization Pouches, GS Medical Packaging
Sterilization Wraps
All STERIL-PEEL® products are manufactured from medical grade bleached kraft paper, hydrogen peroxide, rather than chlorine is used as the bleaching agent
STERIL-PEEL® products are printed with dual indicators which change colour in either steam or ethylene oxide. Both indicators conform with the latest ISO 11140 and EN868 regulations. The steam indicator ink will only change in the presence of all necessary parameters – saturated steam, temperature and time.
They are made from virgin pulp, ensuring optimum consistency of the paper sheet, providing superior barrier properties. Lead-Free and Medically-Safe Inks.
Certifications: 510K and CE Compliant.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty