Would you like to log in again?
Sample Vials, Clear, Without Caps, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
60812-2870 60812-1528 60812-1965 60812-2385 60812-1760 60812-1545 60812-1940 60812-2895 60812-1738 60812-2170 60812-1232 60812-1235
89201-790CS 135.84 USD InStock
89201-790 89201-794 89201-782 89201-792 89201-780 89201-798 89201-786 89201-796 89201-802 89201-778 89201-800 89201-788
Sample Vials, Clear, Without Caps, Kimble Chase, DWK Life Sciences
Vials
Screw thread sample vials are manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Confirms to USP Type I and ASTM E438, Class A requirements.
Ordering information: Closures are not included.
Packaging: Vials are packaged in corrugated trays with partitions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty