Would you like to log in again?
Sample Vials, Screw Thread, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
60957C-4 60920D-4 60910-4 60910L-1 60910-3 60957D-11 60910C-6 60810-1528 60912-1940 60910D-2 60910-2 60920D-8 60912D-1 60910-8 60810-1965 60810-2385 60812B-1232 60912D-2 60920D-1 60957C-11 60810-2170 60812B-12 60910-1 60812B-1 60912-1235 60910D-3 60812B-6 60812B-3 60957D-6 60812B-2 60815-1965 60910C-4 60910C-12 60810-1760 60957D-4 60912-1528 60912-2857 60811S-10 60910L-2 60910-6 60910L-12 60957-4 60912-2895 60812B-4 60810-1940 60910D-4 60912D-12 60957-1 60910C-2 60912-1545 60910C-112 60912-1760 60912-1738 60910C-3 60815-1760 60811S-5 60957C-6 60910D-1 60910C-8 60910C-1
66010-560CS 526.47 USD InStock
66010-560 66010-482 66010-243 66010-484 66010-286 66010-564 66010-322 66010-442 66010-445 66010-247 66010-522 66010-249 66010-446 66010-449 66010-448 66010-408 89201-874 89201-878 89201-876 66010-274 66010-276 66010-431 66010-437 66009-816 66012-146 66012-140 66010-270 66009-813 66010-272 66012-142 89201-806 66010-581 89201-804 66010-265 66010-300 66010-267 89201-808 66010-268 89201-776 89201-852 89201-774 89201-850 89201-812 89201-856 89201-810 66010-260 89201-854 66010-251 66010-570 89201-816 66010-452 66010-253 89201-814 66010-451 89201-858 66010-410 66010-255 89201-860 89201-864 89201-862
Sample Vials, Screw Thread, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Vials
General-purpose, clear and amber borosilicate glass vials.
Volumes range from 0.5 to 11 drams.
Certifications: Meets USP Type I and ASTM E438, Type I, Class A and B requirements.
Ordering information: Available with or without screw caps. Caps are packed separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty