Would you like to log in again?
KIMBLE® Sample Vials, Amber, Phenolic Caps, Kimble Chase
Supplier: DWK Life Sciences
KIMBLE®
60911B-12 60911B-1940 60911B-1528 60911B-10 60911B-2 60911B-5 60911B-1738 60911B-1
89201-828CS 318.1 USD InStock
89201-828 89201-826 89201-820 89201-830 89201-824 89201-834 89201-822 89201-832
KIMBLE® Sample Vials, Amber, Phenolic Caps, Kimble Chase
Vials
Vials manufactured from 51 expansion borosilicate amber glass conforming to USP Type I, with attached black phenolic caps and PTFE-faced white rubber liners.
Packaging: Packaged in corrugated trays with partitions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty