Would you like to log in again?
KIMAX® Sample Vials, Borosilicate Glass, with Black Phenolic Caps and White Rubber Liners, DWK Lifesciences
Supplier: DWK Life Sciences
KIMAX®
60911D-12 60911D-5 60911D-2 60911D-1
89201-838CS 252.81 USD InStock
89201-838 89201-836 89201-842 89201-840
KIMAX® Sample Vials, Borosilicate Glass, with Black Phenolic Caps and White Rubber Liners, DWK Lifesciences
Vials
These sample vials have closed top black phenolic closure with white rubber liners, attached.
  • Amber glass vials
Manufactured from 51 expansion borosilicate amber glass conforming to UPS Type I requirements.
Packaging: Packaged in corrugated trays with partitions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty