Would you like to log in again?
Sample Vials, Amber, with Open-Top Screw Cap, DWK Lifesciences
Supplier: DWK Life Sciences
60911P-10 60911S-5
89201-846CS 217.21 USD InStock
89201-846 89201-844
Sample Vials, Amber, with Open-Top Screw Cap, DWK Lifesciences
Vials
Amber screw thread sample vials are manufactured from 51 expansion borosilicate amber glass conforming to USP Type I requirements.
  • Open-top white polypropylene closures
  • PTFE-faced silicone septa attached
89201-844 features 0.005" PTFE-faced septa on 0.120" silicone liner.
89201-846 features 0.0055" PTFE-faced septa on 0.06" silicone liner.
Packaging: Vials packaged in corrugated trays with partitions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty