Would you like to log in again?
KIMAX® Sample Vials, Borosilicate Glass, with Unattached Black Phenolic Caps and PTFE Faced White Rubber Liners, DWK Lifesciences
Supplier: DWK Life Sciences
KIMAX®
60912B-1232 60912B-1 60912B-12 60912B-2
89201-870CS 241.53 USD InStock
89201-870 89201-872 89201-868 89201-866
KIMAX® Sample Vials, Borosilicate Glass, with Unattached Black Phenolic Caps and PTFE Faced White Rubber Liners, DWK Lifesciences
Vials
Amber Vials with attached black phenolic caps and white rubber liners.
  • Closed top black phenolic closure with white rubber liners, attached
  • Manufactured from 51 expansion borosilicate amber glass conforming to UPS Type I requirement
Packaging: Packaged in corrugated trays with partitions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty