Would you like to log in again?
KIMBLE® Open-Top Polypropylene Screw Thread Caps with Septa, DWK Lifesciences
Supplier: DWK Life Sciences
KIMBLE®
73812BK-13425 73813WH-8425 73813BK-13425 73814BK-13425 73813BK-8425 73814WH-8425 73812BL-9425 73812BK-8425 73812WH-8425 73814BK-13425 73812WH-13425 73813BK-9425 73814BL-9425 73812BK-9425 73813BL-9425 73813WH-13425 N73805-24
89201-904CS 871.76 USD InStock
89201-904 89201-914 89201-902 89201-908 93001-090 89201-906 89201-892 89201-890 89201-886 10061-466 89201-896 89201-894 89201-912 89201-900 89201-888 89201-910 89201-898
KIMBLE® Open-Top Polypropylene Screw Thread Caps with Septa, DWK Lifesciences
Closures Test/Sample Tube Closures
Convenient pre-assembled caps and liners reduce the risk of contamination which makes this to use in autosamplers.
  • Intended for both single and multiple use injection
  • Ideal for storage of volatile compounds and chemicals
  • Autoclavable
Screw thread caps with bonded white PTFE septa available only as standard (no slit), whereas caps with red PTFE faced silicone septa available as both standard and pre slit.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty