Would you like to log in again?
Concept Hand Crank Modular Workstations
Supplier: Treston
10135033 10135032 10135031
89202-094EA 2774.97 USD InStock
89202-094 89202-096 89202-092
Concept Hand Crank Modular Workstations
Furniture Tables and Workbenches
Concept lab benches are particularly suitable for the needs of laboratory industries.
  • Concept hand crank frames can support up to 1100 lbs. static, or 440 lbs. dynamic
  • The hand crank can be installed on the left or right side of the frame and folds away when not in use
  • Upright frames can be added to allow for use of accessories
  • Height is adjustable from 265/6 to 403/5 inches
There are many possible adjustments, all components fit together seamlessly, and each detail is practical in use. The Concept lab bench adapts to the work process and the needs of the user. As a result all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. Concept lab benches are also available in motorized and friction versions.
Ordering information: Work surfaces sold separately. Corner, extension, and double-sided table add on options available. Installation is required. Assembly videos available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty