Would you like to log in again?
Nunc® Cryobank and Bank-It™ Cryogenic Storage Vials, Thermo Scientific
Supplier: Thermo Scientific
Nunc® Bank-It™
374260 374088 374260 374027 374088 374086 374110 374261 374026 374074 374130 374080 374081 374025 374074 374261 374259 374080 374259 374086 374078 374258 374078 374258
82030-514CS 1738.8 USD InStock
82030-514 CA89202-190 89202-192 82030-526 CA82030-522 CA82030-510 CA89202-192 82030-510 89202-186 82030-522 89238-108 89202-188 CA89202-186 CA89202-188 89094-363 89094-365 89094-361 89238-102 89238-104 CA82030-518 82030-518 CA82030-526 89202-190 CA82030-514
Nunc® Cryobank and Bank-It™ Cryogenic Storage Vials, Thermo Scientific
Vials
Vials are designed for low-temperature storage of biological materials.
  • Features low protein adsorption, low DNA adsorption
  • Ideal for cells and proteins, storage of DNA
  • Sterility Assurance Level (SAL) of 10⁻⁶
  • Pyrogen-free according to the LAL-test
Vials are also available with unique 14×14 array 2D coding. The 2D code is laser-etched into the polypropylene bottom of the vial and cannot be lost during storage. The cryogenic storage box (82030-530) is constructed of white cardboard and holds 169 vials in a 13×13 format.
Vials snap into a 96-place automation-compatible rack with a standard barcode to allow for automated manipulation and transport without the risk of losing samples. Vials are also available in loose bulk cases. Caps are internally threaded with a silicone gasket and feature a socket to allow opening and closing with a screwdriver. The dense storage format saves freezer space. Vials withstand temperatures from –196 to 121 °C (–321 to 250 °F).
Certifications: All vials are DNAse- and RNAse-free and are nontoxic according to USP class VI test. Vials conform to IATA requirements for the transport of diagnostic specimens and UN packing instruction 602 and 650. CE marked for diagnostic use.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty