Would you like to log in again?
LABJACK ANODIZED ALUM 15X15
Supplier: Chemglass
Labjack^ Anodized aluminum, 15 x 15cm (6 x 6in) top plate, 18cm (7in) vertical lift
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LABJACK ANODIZED ALUM 15X15 89202-278 CG-3054-10 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty