Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
QuadMag Genie® Four-Position Magnetic Stirrers, 230 V
Supplier: Scientific Industries
SI-5246
89202-328EA 2073.55 USD InStock
89202-328
QuadMag Genie® Four-Position Magnetic Stirrers, 230 V
Stirrers Magnetic Stirrers
These four-position magnetic stirrers are available in an analog model or digital model.
  • 230 V model
Choose the analog model, which is optimized for high speed, or choose the digital model for better speed control; this model offers digital display, control of speed, and is time-speed adjustable in 10 rpm increments. Digital models also incorporate a timer from 1 to 99 minutes and a ramping feature that slowly increases speed to avoid splashing and improve spin bar control. Can be used in cold rooms and incubators.
Powerful, general purpose four-position magnetic stirrers feature "rare earth" magnets that stay coupled under the most demanding strirring conditions and offer exceptional magnetic force capable of stirring up to 2 L for each position
Certifications: CE marked.
Ordering information: 230V models are supplied without a plug.
Caution: 220 V model. Corresponding model with voltage suitable for use in US (89202-326) is also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty