Would you like to log in again?
ECO Immersion Thermostats, LAUDA-Brinkmann
Supplier: Lauda-Brinkmann
L001079 L001080 L001081 L001078
89202-342EA 4420.89 USD InStock
89202-342 89202-344 89202-434 89202-436
ECO Immersion Thermostats, LAUDA-Brinkmann
Circulating Thermostats Open Circulator Systems Thermostat Controllers
ECO current replacement for Ecoline
The intuitive menu navigation allows for easy operation. Both control heads are equipped with a mini-USB interface as a standard feature. Additional interfaces are available via the add-on modules. On-the-fly adjustment of the flow between internal and external circulation is made possible even during operation via the new flow rate switch. The more powerful units are equipped with the energy-saving LAUDA SmartCool system. All cooling thermostats are available with air-cooled or water-cooled options.
There are innovations and developments particularly with regard to the range of features and the ease of operation. Both control heads, designated ECO Silver and ECO Gold, have a powerful circulating pump with more than 30 percent higher pump capacity compared to the predecessor models.
Certifications: CE Certified
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty