Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Talon® Connectors
Supplier: VWR
VWR® Talon®
80862641 915862
89202-616EA 54.6 USD InStock
89202-616 60079-010
VWR® Talon® Connectors
Support Rods and Frames
Use this rod end connector when semi-permanent installations are required.
  • electropolished stainless steel or nickel-plated zinc
  • for rod ends
  • Sturdy
  • Precision bored for close fit with rods
Holds rods firmly at 90°. Comes with two set screws and is precision bored for close fit with 13mm (1/2") rods.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty