Would you like to log in again?
ValuClean® Plus 50% Rayon / 50% Polyester Spunlace Cleanroom Wiper, Berkshire
Supplier: Berkshire
VCLP.0707.45 VCLP.0909.20
89202-694CS 656.63 USD InStock
89202-694 89204-836
ValuClean® Plus 50% Rayon / 50% Polyester Spunlace Cleanroom Wiper, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
ValuClean® Plus is a hydroentangled rayon/polyester nonwoven wiper recommended for ISO class 5 and above environments and offers durability and cleanliness.
Pre wetted option: The same wiper material can be provided in pre-wetted formats for reduced VOC emissions, increased convenience, increased productivity, improved solvent control and cleaning protocol repeatability and reduced costs. Sterile Validated Option: For aseptic processing areas, the same wiper material can be provided in a gamma irradiated validated sterile to a 10⁻⁶ sterility assurance level. Dry and pre-wetted versions available.
Low basis weight reduces landfill waste impact when compared with market leading cellulose/polyester wipers. General wiping in component prep, compounding and wash areas, applying and removing cleaning and disinfecting solutions and spill pick up, general wipe downs, and cleaning of lab equipment.
Due to 50% rayon content, ValuClean® Plus has absorbency reducing actual wiper usage in spill removal applications. Its soft hand and low fiber shedding performance make it a good choice in critical cleaning applications
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty