Would you like to log in again?
VWR® Refrigerated Circulating Baths, 240 V
Supplier: VWR
AD28R-30-V12 AD07R-40-V12 AP45R-20-V12 AP07R-20-V12 AD7LR-20-V12 AP15R-30-V12 AP45R-20-V13D AD07R-20-V12 AD20R-30-V12 AD15R-30-V12 AD15R-40-V12 MX07R-20-V12
89202-968EA 4347.73 USD InStock
89202-968 89203-004 89202-980 89203-024 89203-012 89203-022 89202-992 89202-972 89202-964 89202-996 89202-988 89202-976
VWR® Refrigerated Circulating Baths, 240 V
Circulating Thermostats Closed Circulator Systems Cooling Circulators
VWR® Refrigerated Circulating Baths offer precise temperature control, at temperature ranges from -40 to 200 °C (depending on the controller).
  • Easy-to-view LCD display
  • Precise temperature control
Available in a wide range of sizes, with three controller options, these refrigerated circulating baths are ideal for a variety of applications.
The large LCD display on the MX temperature controller is easily viewed across the laboratory. The Advanced Digital temperature controller offers a touch-pad, while the Advanced Programmable Temperature Controller includes a full touch-screen display, and programmability (ten 100-step programs).
The Advanced Series Controllers offer Swivel 180™ technology, which allows you to rotate the controller independently from the bath. All heated circulating baths include LidDock™ lid stowing system and DuraTop™, the precision molded phenolic bath top, which remains cooler at high temperatures and resists laboratory chemicals.
Certifications: ETL listed. CE marked. MX Controller complies with DIN 12876-1 Class I safety requirements for use with non-flammable liquids. Advanced Digital and Advanced Programmable comply with DIN 12876-1 Class III safety requirements for use with flammable liquids. cETLus compliant.
Ordering information: Refrigerated Circulators include reservoir cover; bypass tubing; male inlet and outlet adapters for 3/8", 1/4" and 3/16" (.95, .63 and .47 cm) tubing. 1/4" NPT to M16 adapters also included on 50 Hz models.
Caution: Note: These units are all 240 V, 50Hz units. Corresponding 120 V models are also available, these are 89202-962, 89202-966, 89202-970, 89202-974, 89202-978, 89202-986, 89202-990, 89202-994, 89203-002, 89203-010 and 89203-014 respectively.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty