Particle-free, residue-free tapes.
- High adhesion
- Residue-free
- Easily marked with a pen
- Wide assortment of colors and sizes
UltraTape's polyethylene tape is ideal for any cleanroom construction application that requires a strong bond. This high adhesion, polyethylene tape is residue-free, easy to tear, and can be written on with a pen. A uniquely striped color pattern allows immediate recognition of the product as a certified cleanroom tape.