Would you like to log in again?
REFRACTOMETER, MULTI-CHEK F
Supplier: Reichert Technologies
Reichert Multi-Chek (F)^ Is A 4 Scale Digital Pocket Refractometer That Measures Ethylene Glycol Coolant freeze point, propylene glycol coolant freeze point, battery electrolyte specific gravity, and windshield washer fluid freezepoint
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REFRACTOMETER, MULTI-CHEK F 89203-922 13940014 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty