Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PURELAB® Classic Water Purification Systems, ELGA LabWater
Supplier: ELGA LabWater
PURELAB® Classic
CLXXXUFM2 CLXXXUVM2 SKCSPCLASSICUV CLXXUVFM2 CLXXXDIM2 CLXXXDIM2-US SK-CSPCLASSICA CLXXUVFM2-US CLXXXUFM2-US SK-CSPCLASSICB CLXXXUVM2-US
76516-392KT 1791.14 USD InStock
76516-392 CA89204-076 76516-394 76516-396 CA89204-072 89204-084 89204-076 CA89204-084 89204-080 89204-072 CA89204-080
PURELAB® Classic Water Purification Systems, ELGA LabWater
Water Purification Systems Pure and Ultra Pure Water Systems
The PURELAB Classic system combines high performance with economy to deliver 18.2 MΩ-cm type I ultrapure water at a very cost-effective price
  • Ultrapure type I water at very economic costs for the equipment and the cost of ownership
  • Automatic intermittent recirculation minimizes temperature build-up and optimizes microbial performance
  • Very easy to maintain – incorporates ‘fast rinse’ ultra filter
  • Upgradable from single pack to twin pack purification
  • Suitable for all life science and analytical applications
  • Complete sanitization of all wetted parts ensures optimum microbial performance
The PURELAB Classic contains many market leading features to deliver consistent and reliable water purity.
Accessories information: Additional accessories are available; contact your Avantor representative for more information.
Ordering information: Feed water must be from a pre-treated source such as deionized, distilled or reverse osmosis. If the feed pressure exceeds 10 psi, a pressure regulator is required. One purification pack required and sold separately and if required, a point of use filter can be purchased. Validation labor not included with systems that include validation documentation.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: ELGA LabWater provides several service options such as Extended Warranty, Installation, Preventative Maintenance, and Validation. To learn more about these services please search "Elga Water Technical Service" in the website search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST@vwr.com for further details.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty