Would you like to log in again?
LIQUIPORT™ NF300 and NF1.300 Series Diaphragm Liquid Pumps, KNF
Supplier: KNF
LIQUIPORT®
179112/153257 179106/153257 179107/153257 179104/153257 179108/153257 179103/153257 179110/153257 179111/153257 179102/153257 179113/153257 179105/153257 179109/153257
89204-522EA 2123.85 USD InStock
89204-522 89204-500 89204-524 89204-502 89204-496 89204-520 89204-498 89204-492 89204-518 89204-494 89204-526 89204-516
LIQUIPORT™ NF300 and NF1.300 Series Diaphragm Liquid Pumps, KNF
Pumps Diaphragm Pumps
Chemically resistant liquid transfer diaphragm pump. Offers wetted material combinations for optimal chemical resistance. Manual control of flow rate with LED indicator. For convenient transfer and dosing of neutral or corrosive liquids. Option for analog and impulse control (RC).
  • Premium corrosion resistance
  • Manual front-panel flowrate control
  • Pressure limit protection
  • No metal wetted parts to corrode
  • Reliable splash-proof housing
  • Models are offered with either PP, PVDF or PTFE pump head material depending on the liquid being transferred
  • All models feature PTFE diaphragms and FFPM valves
  • NF300 has a maximum pressure of 1 bar and NF1.300 has a maximum pressure of 4 bar
Pumps are self-priming, maintenance-free and will operate dry indefinitely without damage. Select the best pump based on the desired flowrate, maximum outlet pressure and the wetted materials.
Three considerations for any liquid diaphragm application: The inlet of pump should not be lower than the level of liquid being transferred. An inlet filter is required if media contains particulates or fibers larger than 50 µm. Pulsation varies between specific pump models.
Flow rate: 0.5 to 3 L/min
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty