Would you like to log in again?
Ferrule Removal Kit, Restek
Supplier: Restek
20146
89205-668EA 93.13 USD InStock
89205-668
Ferrule Removal Kit, Restek
Puller and Inserter Tools
The tapered tools in this kit have teeth designed to grip and remove fused silica adapter ferrules that have become stuck in the fitting detail.
Each kit has two tools: one for removing 1/32-inch adapter ferrules and one for removing 1/16-inch adapter ferrules.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty