Would you like to log in again?
Spill-X-A® Acid-Neutralizing Adsorbent, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GEN381 GEN322
89205-854EA 766.01 USD InStock
89205-854 89205-848
Spill-X-A® Acid-Neutralizing Adsorbent, New Pig
Spill Control Materials
Neutralizes and solidifies most mineral and organic acids for safe and easy cleanup, including sulfuric, hydrochloric, nitric, and hydrofluoric
Granular base lets you use only what you need; reduces product waste and disposal costs. Speeds spill cleanup to limit worker exposure and minimize production downtime. Excellent for use in shipping/receiving areas, acid container storage areas and acid use and transfer sites. Non-toxic, free-flowing dry chemical is environmentally friendly.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty