Would you like to log in again?
Spill-X-C® Caustic-Neutralizing Adsorbent, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GEN323 GEN382
89205-860EA 1224.61 USD InStock
89205-860 89205-850
Spill-X-C® Caustic-Neutralizing Adsorbent, New Pig
Spill Control Materials
Neutralizes and solidifies many caustics for safe and easy cleanup, including sodium hydroxide (50%) and ammonium hydroxide (29%)
Granular base lets you use only what you need; reduces product waste and disposal costs. Speeds spill cleanup to limit worker exposure and minimize production downtime. Excellent for use in shipping/receiving areas, caustic container storage areas and caustic use and transfer sites. Non-toxic, free-flowing dry chemical is environmentally friendly.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty