Would you like to log in again?
Orion™ Versa Star Pro™ Benchtop Meter without Modules, Thermo Scientific
Supplier: Thermo Scientific
Orion™ Versa Star Pro™
VSTAR00
89206-376EA 1587.22 USD InStock
89206-376
Orion™ Versa Star Pro™ Benchtop Meter without Modules, Thermo Scientific
Electrochemical Meters Conductivity Meters
Designed to provide optimal flexibility, the Orion™ Versa Star Pro™ Benchtop Meter without Modules can be combined with up to four measurement modules to build an ideal system.
  • Onscreen measurement stability indicator lets users know exactly when measurements are ready
  • Auto-Read mode holds a stable reading, Continuous mode shows changing readings, Timed mode logs data at specific intervals, and Single-Shot mode holds a reading after a preset time is reached
  • Audible alarms automatically notify users when a set offset value, high/low limit value, and calibration time interval are reached
  • Easily access, update, and switch preferred measurement settings, using ten password-protected methods per channel
  • Uniformly mix solutions using up to two meter-controlled stirrer probes
  • Ensure measurement integrity by viewing the active calibration log while taking readings
  • Three-year meter warranty
For the ultimate flexibility, combine the Orion™ Versa Star Pro™ Benchtop Meter without Modules with up to four Orion™ Versa Star™ measurement modules to build an ideal electrochemistry measurement system.
Orion™ Versa Star Pro™ benchtop meters are designed to meet the most challenging applications with ease. Delivering rich functionality and features, users can optimally configure the meter system to meet changing requirements. These meters offer interchangeable measurement modules that allow multiple users to customize four separate channels to meet their specific requirements. Easily view measurements on a stunning color display with adjustable text sizes. Take the guesswork out of meter calibration and setup with onscreen instructions available in a variety of local languages. Enjoy quick and simple data entry using the numeric keypad. Data log collects and date/time stamps up to 2000 measurement sets with optional electrode ID, sample ID, and account ID.
Meter facilitates data transfers to a computer via USB or RS232 connections. Non-volatile meter memory preserves data and settings, even in the event of power loss. IP54-rated housing resists dust and splashes, and can be wall-mounted.
Certifications: CE, TUV 3-1, FCC Class AIP-54.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty