Would you like to log in again?
Orion™ Star™ A Series Benchtop Meter-Attached Electrode Stand, Thermo Scientific
Supplier: Thermo Scientific
Orion™
STARA-BEA
89206-426EA 299.83 USD InStock
89206-426
Orion™ Star™ A Series Benchtop Meter-Attached Electrode Stand, Thermo Scientific
Electrode Holders
This meter-attached electrode stand can be used to easily place sensors in samples.
  • Simplifies rinsing and storage between samples
  • Cable management system minimizes electrode breakage
  • 360° motion allows electrodes to be moved vertically, horizontally, and front-to-back positioning into beakers or sample containers
Can be attached to either side of Orion™ Versa Star benchtop meters and Orion™ Star™ A210 and Star™ A110 series benchtop meters. Can also be used with weighted base for compatibility with any meter system (portable or benchtop).
Ordering information: Includes meter bracket, arm, and electrode holder. The weighted base must be ordered separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty