Would you like to log in again?
BIN STAT DISP BEN 9X7X12 PK12
Supplier: Metro
Olympic* Stacking Bin Blue 37.5 cm^ Made of Benstat* static-dissipative polypropylene-base material. Durable, stackable, with high impact strength. Surface resistivity of more than 1e5 ohms/sq. and less than 1e12 ohms/sq. Exterior dimensions: 21wx37.5lx17.8hcm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BIN STAT DISP BEN 9X7X12 PK12 89207-072 SB91587BAS Each Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty