Would you like to log in again?
Whatman™ Cellulose Blotting Papers, Grade GB003, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
10427813 10426892 10427806 10427813 10427805 10426880 10547922 10426890 10427805 10547922 10427818 10427804 10427804 10427812 10427810 10427826 10427810 10426880 10427812 10426892 10426890 10427826
89026-868PK 268.03 USD InStock
CA89026-868 89026-868 CA89207-199 CA89207-200 89207-200 CA74330-134 89207-198 CA28153-293 28153-293 89026-870 89207-204 74330-134 CA89207-204 52844-508 52844-509 28152-033 CA89026-872 CA52844-508 28152-032 CA89026-870 89026-872 CA52844-509
Whatman™ Cellulose Blotting Papers, Grade GB003, Whatman products (Cytiva)
Blotting Paper
This general purpose blotting paper is made from pure raw materials with a high absorbency and is used as a membrane gel support. Due to its thickness (0,8 mm), it is recommended for the lysis/denaturation of colony or plaque lifts and Western blots.
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives ensures that no contamination will occur during the transfer steps
  • Manufactured and tested specifically for chromatographic and blotting techniques ensuring the wicking capability and uniformity of capillary action that are important in obtaining clean and even transfers during blotting
  • Industry standard for blotting procedures
  • Convenient sizes available in sheets precisely cut to the most popular gel and transfer membrane sizes allows 'out-of-the-box' usage and eliminates sheet-to-sheet variation
A thick paper recommended for the lysis/denaturation of colony or plaque lifts and Western blots.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty