Would you like to log in again?
Catheter Procedure Carts, Eagle MHC
Supplier: Eagle MHC
CPC6024 CPC6036D CPC6024D CPC6036
89207-548EA 3866.54 USD InStock
89207-548 89207-546 89207-544 89207-550
Catheter Procedure Carts, Eagle MHC
Carts
Combination of hooks, shelves, and tote boxes give the versatility to hold all sizes and styles of catheter packages.
  • Bars are readily removable
  • Deluxe models come standard with hooks, shelves and tote boxes, and all accessories
  • Hooks, shelf dividers, and tote boxes allow for easy organization
  • Label holders are provided to help in the organization of inventory
Adjustable components. Shelves are vertically adjustable on one-inch increments, while the hook and bar assemblies can be adjusted to accommodate peel pouches 3" (76 mm) or greater in width.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty