Would you like to log in again?
OPTI-LYNX™ Guard Column Single Cartridges
Supplier: Optimize Technologies
OPTI-LYNX™
11-02255-EB 11-02308-EH 11-02640-EA 11-02255-EE 11-02640-EC 11-02640-JD 11-02255-EH 11-02640-EH 11-02308-EG 11-02308-ED 11-02308-EF 11-02255-JD 11-02308-EB 11-02640-EG 11-02308-EA 11-02308-JD 11-02255-EA 11-02255-EC 11-02255-EF 11-02640-EB 11-02640-ED 11-02640-EF 11-02255-EG 11-02308-EC 11-02255-ED 11-02640-EE 11-02255-MB 11-02308-EE
83008-170EA 227.93 USD InStock
83008-170 89207-696 89207-694 89207-692 89207-690 77100-334 89207-704 77100-336 89207-702 89207-724 89207-722 89207-700 89207-688 89207-720 77100-330 89207-708 77100-332 89207-706 89207-716 77100-324 89207-714 77100-326 89207-712 77100-328 89207-710 89207-698 89207-718 77100-322
OPTI-LYNX™ Guard Column Single Cartridges
Chromatography Columns
The OPTI-LYNX™ Guard Column is a high performance, cartridge-based guard system, suitable for use with any analytical column.
  • Can be used with any brand of column
  • Cartridge based system
  • Low dispersion cartridge interface
  • ZDV holder connection
  • Less than 2 µL swept volume
  • Extends column life
  • Provides effective protection with negligible impact
  • Rated to 6,000 psi
Some cartridge-based guard column products offer convenience with a price: higher internal volumes. At Optimize Technologies, we refuse to compromise performance for anything. OPTI-LYNX™ utilizes low dispersion component interfaces to keep extra column volume to a minimum: connection paths have a total internal volume of less than 2 µl. You get quick-connect convenience and low dispersion, all in one package.
Since the interacting surfaces are identical, the only variable component is the holder tube. Install a guard column for one application and then switch to a pre-column filter for the next. Use an in-line filter with different porosities and diameters just by changing the insert.
Ordering information: Available individually and in packs of five. Custom packing available.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty