Would you like to log in again?
Milk Dilution Bottles
Supplier: Corning
70368-160 70373-160
89208-248CS 570.66 USD InStock
89208-248 89208-252
Milk Dilution Bottles
Bottles Dilution Bottles
Made of Corning soda lime glass.
  • Wide mouth bottle for easy measuring or filling
  • Uses a GPI 40-400 threaded phenolic screw cap with rubber liner
  • Autoclaving at 121 °C
Used for general storage of solutions and tissue culture work.
Certifications: Meets the requirements for milk dilution bottles stated in the 'Standard Methods for the Examination of Dairy Products', published by the American Public Health Association.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty