Would you like to log in again?
Bulk Dispenser with Lid, S-Curve Technologies
Supplier: S-Curve Technologies
BD-03 L BD-02 L
89208-614EA 452.06 USD InStock
89208-614 89208-612
Bulk Dispenser with Lid, S-Curve Technologies
PPE Dispensers
2 & 3 compartment dispensers with lid for bulk items (gloves, face masks, hair nets, bouffants, shoe covers, etc.).
  • Available in 1/4″ clear acrylic or 1/4″ clear PETG material – safe with IPA
  • Open top for easy access to product
  • Flame treated edges for smoothness
Rubber pads on bottom corners to prevent slipping & scratching
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty