Would you like to log in again?
Dispenser with 8 Compartments, S-Curve Technologies
Supplier: S-Curve Technologies
BD-5300 BD-5300 PETG
89208-624EA 341.97 USD InStock
89208-624 89208-626
Dispenser with 8 Compartments, S-Curve Technologies
PPE Dispensers
Eight compartments to organize and store small items.
  • 1/4″ clear acrylic or 1/4″ clear PETG material
  • Open top for easy access to product
  • Flame polished edges for a smooth finish
  • Keyhole standoffs on back for wall mounting
Wall mountable: no.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty