Would you like to log in again?
VWR® Advanced Disposable Sleeve
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
89208-810
89208-810CS 284.27 USD InStock
89208-810
VWR® Advanced Disposable Sleeve
Disposable Garments
Sleeves provide reliable protection in laboratory, electrical, pharmaceutical, food, industrial, and cleanroom applications.
  • Disposable fabrication
  • Reliable protection in a variety of applications
Disposable non-woven sleeve is made of 56gsm SF fabric. The Advanced Protection sleeve features a knit cuff one at end, and sonic-welded seams.
Certifications: ISO9001
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty