Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Jacketed Filter Reactor Systems, 10 L, Electric Motor, Chemglass

Supplier: Chemglass
CG-1959-F610
89209-352EA 64009.87 USD InStock
89209-352
Jacketed Filter Reactor Systems, 10 L, Electric Motor, Chemglass
Laboratory Reactor Systems
Jacketed filter glass reactors feature single or multi-step.

  • Complete reactor system on open air, auto center stands
  • Maximum temperature of 150 °C limit for filter base
  • Filter base eliminates material transfers and significantly reduces losses

Can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis.

Ordering information: Filter reactors are for vacuum/gravity filtration use only.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty