Would you like to log in again?
FHP NanoTech Microfiber Cloth, Vileda Professional-FHP
Supplier: Vileda Professional
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
128601 128600 128603 128602
89209-466CS 661.83 USD InStock
89209-466 89209-460 89209-464 89209-462
FHP NanoTech Microfiber Cloth, Vileda Professional-FHP
Wipes Reusable Cleaning Cloths Cloths for General Applications
NanoTech Micro is the solution for all operations and environments where maximum touch area hygiene is paramount.
  • Nano silver particles are embedded in the endless microfibers
  • Proven cleaning efficacy
  • No growth of bacterial odors or germs on cloths for more than 24 hours
  • Extremely low linting cloths
  • Omit tumble drying, as cloths can be stored in moist conditions for 24 hours without bacteria odor growth
  • Maintains antibacterial properties after 300+ laundry cycles
  • Color-coded to prevent cross contamination
The NanoTech microfiber cloth's streak-free performance provides one-step cleaning with no drying cycle. The cloths can be used pre-prepared or via spray bottle method, giving users a variety of cleaning methods. Different color options reduce cross contamination. Reduce cost and eliminate odor with minimal chemical use.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty