Would you like to log in again?
OPTI-PAK® Trap Columns, 5 µl
Supplier: Optimize Technologies
OPTI-PAK®
10-03233-TE 10-03233-TA 10-03233-TD 10-03233-TF 10-03233-TG 10-03233-TB 10-03233-TC
89209-666PK 1578.94 USD InStock
89209-666 89209-654 89209-664 89209-658 89209-656 89209-662 89209-660
OPTI-PAK® Trap Columns, 5 µl
Chromatography Columns
The OPTI-PAK®'s unique hardware design integrates a PEEK holder with an auto-adjusting stem which provides a fluid interface with any 10-32 standard injector port, and guarantees a zero-dead-volume connection despite variances in tube stop depth.
  • Specially designed for use in today's low volume trapping applications
  • Packed stem is positioned directly in flow path
  • Absolute minimum sample dilution and dispersion
  • Auto adjusting ZDV connection
  • Amenable to any trapping technique
  • Can be installed side-by-side in same valve stator without interference
A proprietary method of frit placement allows the packed stem to be positioned directly into the flow path, which eliminates all excess swept volume. Less swept volume translates to absolute minimum sample dilution and dispersion. The elegant, flexible design of OPTI-PAK makes it amenable to virtually any trapping application. Installing an OPTI-PAK into the outlet of a switching valve will facilitate the trapping, desalting, and pre-concentration of an analyte mixture. With OPTI-PAK's versatility, a second trap can be added downstream in the loop of an injection valve and effectively perform a 2D separation.
Delivery information: Includes five disposable 10-32 hand-tight holders and custom packed cartridges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty