Would you like to log in again?
Snap Caps for Blood Drawing Tubes, Globe Scientific
Supplier: Globe Scientific
113148C 113146C
89209-806PK 43.94 USD InStock
89209-806 89209-808
Snap Caps for Blood Drawing Tubes, Globe Scientific
Closures Test/Sample Tube Closures
Snap-on closures are an effective way to recap blood drawing tubes, glass test tubes, and plastic culture tubes
They are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation, and refrigeration procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty